Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCCGTAGGGTGGGCTGGAAGCT[A/G]GGAACTGACTGAAAACATTGAGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB37 - RAB37, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006638.2 | Intron | NP_001006639.1 | ||||
NM_001163989.1 | Intron | NP_001157461.1 | ||||
NM_001163990.1 | Intron | NP_001157462.1 | ||||
NM_175738.4 | Intron | NP_783865.1 | ||||
XM_005257288.4 | Intron | XP_005257345.1 |