Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGCGGGCGCTCTTGCGCCCATC[C/T]TCGGAGACCTTGAAGGTGCCTTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607827 MIM: 607696 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTOP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTOP2 - otopetrin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USH1G - USH1 protein network component sans | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282489.2 | 868 | Silent Mutation | GAA,GAG | E,E 132 | NP_001269418.1 | |
NM_173477.4 | 868 | Silent Mutation | GAA,GAG | E,E 235 | NP_775748.2 | |
XM_011524296.2 | 868 | Silent Mutation | GAA,GAG | E,E 132 | XP_011522598.1 |