Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGCTGTGGAACGTGAAGAAGGA[A/G]GTAAAGTCGAGTCAAGAACCCCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611418 MIM: 611214 | ||||||||||||||||||||
Literature Links: |
SGSM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SGSM2 - small G protein signaling modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098509.1 | 172 | Silent Mutation | GAA,GAG | E,E 19 | NP_001091979.1 | |
NM_014853.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | NP_055668.2 | |
XM_011524101.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522403.1 | |
XM_011524102.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522404.1 | |
XM_011524103.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522405.1 | |
XM_011524104.2 | 172 | Intron | XP_011522406.1 | |||
XM_011524105.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522407.1 | |
XM_011524106.1 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522408.1 | |
XM_011524107.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522409.1 | |
XM_011524108.1 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522410.1 | |
XM_011524109.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522411.1 | |
XM_011524110.2 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_011522412.1 | |
XM_017025474.1 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_016880963.1 | |
XM_017025475.1 | 172 | Silent Mutation | GAA,GAG | E,E 19 | XP_016880964.1 | |
XM_017025476.1 | 172 | Intron | XP_016880965.1 |
SNORD91A - small nucleolar RNA, C/D box 91A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD91B - small nucleolar RNA, C/D box 91B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSR1 - TSR1, ribosome maturation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018128.4 | 172 | Intron | NP_060598.3 |