Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAGACTTAAAAGCACATAGCTA[C/T]GTGGCAAAACTTTTTCTTCTTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606699 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HELZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HELZ - helicase with zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014877.3 | 7969 | UTR 3 | NP_055692.2 | |||
XM_005257888.4 | 7969 | UTR 3 | XP_005257945.1 | |||
XM_005257889.4 | 7969 | UTR 3 | XP_005257946.1 | |||
XM_006722214.3 | 7969 | UTR 3 | XP_006722277.1 | |||
XM_006722215.3 | 7969 | UTR 3 | XP_006722278.1 | |||
XM_006722216.3 | 7969 | UTR 3 | XP_006722279.1 | |||
XM_011525544.2 | 7969 | UTR 3 | XP_011523846.1 | |||
XM_017025477.1 | 7969 | UTR 3 | XP_016880966.1 | |||
XM_017025478.1 | 7969 | UTR 3 | XP_016880967.1 |