Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTCAGTGTGCTGTCACTGCTGC[C/T]GCTGACCAGGAGCCGACTGTCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNDC8 - fibronectin type III domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NLE1 - notchless homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014445.2 | 1405 | Missense Mutation | NP_001014445.1 | |||
NM_018096.4 | 1405 | Missense Mutation | NP_060566.2 | |||
XM_017024777.1 | 1405 | Missense Mutation | XP_016880266.1 |