Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCACCGTCCCTGAGATGCCACCA[A/G]CCACAAAGCTCATGCCCACAGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605448 MIM: 610820 | ||||||||||||||||||||
Literature Links: |
RUNDC3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RUNDC3A - RUN domain containing 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A39 - solute carrier family 25 member 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143780.2 | 872 | Missense Mutation | GCT,GTT | A,V 261 | NP_001137252.1 | |
NM_001321240.1 | 872 | Missense Mutation | GCT,GTT | A,V 238 | NP_001308169.1 | |
NM_001321241.1 | 872 | Missense Mutation | GCT,GTT | A,V 259 | NP_001308170.1 | |
NM_016016.3 | 872 | Missense Mutation | GCT,GTT | A,V 253 | NP_057100.1 | |
XM_011524880.2 | 872 | Missense Mutation | GCT,GTT | A,V 267 | XP_011523182.1 | |
XM_011524882.2 | 872 | Missense Mutation | GCT,GTT | A,V 244 | XP_011523184.1 | |
XM_011524883.2 | 872 | Missense Mutation | GCT,GTT | A,V 166 | XP_011523185.1 | |
XM_011524884.1 | 872 | Missense Mutation | GCT,GTT | A,V 166 | XP_011523186.1 | |
XM_017024737.1 | 872 | Missense Mutation | GCT,GTT | A,V 160 | XP_016880226.1 |