Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTGCAGCCCAGGAAGAAGACGC[C/G]GAAGACCACGGTAAGCGACAGCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604027 | ||||||||||||||||||||
Literature Links: |
GOSR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GOSR2 - golgi SNAP receptor complex member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5089 - microRNA 5089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPRML - reprimo like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203400.4 | 488 | Missense Mutation | CGC,GGC | R,G 83 | NP_981945.1 |