Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACGCCCATAGGAGAAGGTAAGC[A/G]CCCAGGGTCGGGGAAGGGGGCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103870 MIM: 610271 MIM: 615562 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDOC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDOC - aldolase, fructose-bisphosphate C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005165.2 | 1035 | Missense Mutation | GCG,GTG | A,V 297 | NP_005156.1 | |
XM_005257949.2 | 1035 | Missense Mutation | GCG,GTG | A,V 297 | XP_005258006.1 | |
XM_011524556.1 | 1035 | Missense Mutation | GCG,GTG | A,V 297 | XP_011522858.1 |
PIGS - phosphatidylinositol glycan anchor biosynthesis class S | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPAG5 - sperm associated antigen 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |