Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGTTACCCTCCTTCTTTCCCAG[C/G]TCCTGAAACTGGGCACTGGTCTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607763 MIM: 609848 | ||||||||||||||||||||
Literature Links: |
ACAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACAP1 - ArfGAP with coiled-coil, ankyrin repeat and PH domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014716.3 | 318 | Missense Mutation | CTC,GTC | L,V 38 | NP_055531.1 |
KCTD11 - potassium channel tetramerization domain containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |