Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTTCCCTCCCTCTCTCATTCAT[C/T]CATTTTCCTCCCTCCCCTCCAGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118820 MIM: 139240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSH2 - chorionic somatomammotropin hormone 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GH2 - growth hormone 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002059.4 | 783 | Intron | NP_002050.1 | |||
NM_022556.3 | 783 | Intron | NP_072050.1 | |||
NM_022557.3 | 783 | Nonsense Mutation | TGA,TGG | *,W 214 | NP_072051.1 | |
NM_022558.3 | 783 | Intron | NP_072052.1 |
LOC107987248 - intercellular adhesion molecule 1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |