Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACACATGTACTCTTTCTTATAA[C/G]ACTGTTAAAGCAGAAACTCAAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606221 MIM: 608499 | ||||||||||||||||||||
Literature Links: |
IKZF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IKZF3 - IKAROS family zinc finger 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZPBP2 - zona pellucida binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198844.2 | 429 | Missense Mutation | AAC,AAG | N,K 92 | NP_942141.2 | |
NM_199321.2 | 429 | Missense Mutation | AAC,AAG | N,K 114 | NP_955353.1 | |
XM_011524298.1 | 429 | Missense Mutation | AAC,AAG | N,K 114 | XP_011522600.1 |