Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCTCTTCTGTATCAGCAGCTCG[C/T]GACAAGATGTCTGCCATCAGTGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603547 | ||||||||||||||||||||
Literature Links: |
MBD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBD2 - methyl-CpG binding domain protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003927.4 | 1408 | Silent Mutation | TCA,TCG | S,S 393 | NP_003918.1 | |
NM_015832.4 | 1408 | Intron | NP_056647.1 |