Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAAGGCATGGTTTTTTCAATTA[C/T]TGATGGGGACAATTTTCCTCTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605252 MIM: 607051 | ||||||||||||||||||||
Literature Links: |
POLI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLI - DNA polymerase iota | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STARD6 - StAR related lipid transfer domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139171.1 | 699 | Missense Mutation | ATA,GTA | I,V 183 | NP_631910.1 | |
XM_011525821.1 | 699 | Missense Mutation | ATA,GTA | I,V 183 | XP_011524123.1 | |
XM_017025552.1 | 699 | Missense Mutation | ATA,GTA | I,V 124 | XP_016881041.1 | |
XM_017025553.1 | 699 | Missense Mutation | ATA,GTA | I,V 124 | XP_016881042.1 | |
XM_017025554.1 | 699 | Missense Mutation | ATA,GTA | I,V 101 | XP_016881043.1 |