Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGAATGAGTCCACCTAGTCTTA[C/T]GTAACAGTGATAAATTAATATTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606930 MIM: 607274 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THOC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THOC1 - THO complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005131.2 | Intron | NP_005122.2 | ||||
XM_011525772.2 | Intron | XP_011524074.1 | ||||
XM_011525773.1 | Intron | XP_011524075.1 | ||||
XM_011525774.2 | Intron | XP_011524076.1 | ||||
XM_017026103.1 | Intron | XP_016881592.1 | ||||
XM_017026104.1 | Intron | XP_016881593.1 |
USP14 - ubiquitin specific peptidase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |