Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTTCCGAGTCTTGCGTTCCCGC[A/T]TTATGGTACTACTCCTCAGCTCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603016 | ||||||||||||||||||||
Literature Links: |
CDH19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH19 - cadherin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271028.1 | 2180 | UTR 3 | NP_001257957.1 | |||
NM_021153.3 | 2180 | Missense Mutation | AAG,ATG | K,M 665 | NP_066976.1 | |
XM_011525931.2 | 2180 | Missense Mutation | AAG,ATG | K,M 454 | XP_011524233.1 | |
XM_011525932.1 | 2180 | Intron | XP_011524234.1 | |||
XM_017025711.1 | 2180 | Missense Mutation | AAG,ATG | K,M 454 | XP_016881200.1 |