Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATACCTTTGTCCTAGGAGTTTCA[C/G]ATGAAAGTGCTACCTGGGTTTTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616909 | ||||||||||||||||||||
Literature Links: |
CCDC68 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC68 - coiled-coil domain containing 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143829.1 | 1184 | Missense Mutation | TCT,TGT | S,C 303 | NP_001137301.1 | |
NM_025214.2 | 1184 | Missense Mutation | TCT,TGT | S,C 303 | NP_079490.1 | |
XM_006722552.3 | 1184 | Missense Mutation | TCT,TGT | S,C 303 | XP_006722615.1 | |
XM_011526198.2 | 1184 | Missense Mutation | TCT,TGT | S,C 303 | XP_011524500.1 | |
XM_011526199.2 | 1184 | Missense Mutation | TCT,TGT | S,C 303 | XP_011524501.1 | |
XM_011526200.2 | 1184 | Missense Mutation | TCT,TGT | S,C 303 | XP_011524502.1 | |
XM_011526201.2 | 1184 | Missense Mutation | TCT,TGT | S,C 303 | XP_011524503.1 | |
XM_011526202.1 | 1184 | Intron | XP_011524504.1 | |||
XM_011526203.2 | 1184 | Missense Mutation | TCT,TGT | S,C 256 | XP_011524505.1 | |
XM_011526204.2 | 1184 | Intron | XP_011524506.1 | |||
XM_017026011.1 | 1184 | Missense Mutation | TCT,TGT | S,C 256 | XP_016881500.1 |