Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGTTTGAGTCCCTCCTGGGCA[C/T]CTGCCACCCTGTGGAACCAGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBLC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBLC - Cbl proto-oncogene C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130852.1 | 619 | Missense Mutation | ACC,ATC | T,I 181 | NP_001124324.1 | |
NM_012116.3 | 619 | Missense Mutation | ACC,ATC | T,I 181 | NP_036248.3 | |
XM_005258696.3 | 619 | Missense Mutation | ACC,ATC | T,I 181 | XP_005258753.1 | |
XM_011526688.2 | 619 | Missense Mutation | ACC,ATC | T,I 181 | XP_011524990.1 | |
XM_011526689.2 | 619 | Missense Mutation | ACC,ATC | T,I 181 | XP_011524991.1 | |
XM_011526690.2 | 619 | Missense Mutation | ACC,ATC | T,I 181 | XP_011524992.1 |