Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATAAATCTAGGCTTTCCACTCC[A/G]TATATTAGCTGGGGCTGCCATAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611024 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF667 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF667 - zinc finger protein 667 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321355.1 | 3723 | UTR 3 | NP_001308284.1 | |||
NM_001321356.1 | 3723 | UTR 3 | NP_001308285.1 | |||
NM_022103.3 | 3723 | UTR 3 | NP_071386.3 | |||
XM_011527208.1 | 3723 | UTR 3 | XP_011525510.1 | |||
XM_011527209.1 | 3723 | UTR 3 | XP_011525511.1 |