Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCTGATGTGCAATGAGCATGCTT[C/T]TCCCAGTTAAGCCTTTGCCACATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605422 | ||||||||||||||||||||
Literature Links: |
ZNF350 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF350 - zinc finger protein 350 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF615 - zinc finger protein 615 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199324.1 | 2089 | Missense Mutation | AAA,AGA | K,R 591 | NP_001186253.1 | |
NM_001321317.1 | 2089 | Missense Mutation | AAA,AGA | K,R 585 | NP_001308246.1 | |
NM_001321318.1 | 2089 | Missense Mutation | AAA,AGA | K,R 587 | NP_001308247.1 | |
NM_001321319.1 | 2089 | Missense Mutation | AAA,AGA | K,R 591 | NP_001308248.1 | |
NM_001321320.1 | 2089 | Missense Mutation | AAA,AGA | K,R 580 | NP_001308249.1 | |
NM_001321321.1 | 2089 | Missense Mutation | AAA,AGA | K,R 591 | NP_001308250.1 | |
NM_001321322.1 | 2089 | Missense Mutation | AAA,AGA | K,R 587 | NP_001308251.1 | |
NM_001321323.1 | 2089 | Missense Mutation | AAA,AGA | K,R 596 | NP_001308252.1 | |
NM_198480.3 | 2089 | Missense Mutation | AAA,AGA | K,R 580 | NP_940882.3 | |
XM_011526824.1 | 2089 | Missense Mutation | AAA,AGA | K,R 591 | XP_011525126.1 | |
XM_011526825.1 | 2089 | Missense Mutation | AAA,AGA | K,R 591 | XP_011525127.1 | |
XM_011526826.1 | 2089 | Missense Mutation | AAA,AGA | K,R 591 | XP_011525128.1 | |
XM_011526829.1 | 2089 | Missense Mutation | AAA,AGA | K,R 472 | XP_011525131.1 | |
XM_011526830.1 | 2089 | Missense Mutation | AAA,AGA | K,R 472 | XP_011525132.1 | |
XM_017026648.1 | 2089 | Missense Mutation | AAA,AGA | K,R 580 | XP_016882137.1 | |
XM_017026649.1 | 2089 | Missense Mutation | AAA,AGA | K,R 580 | XP_016882138.1 | |
XM_017026650.1 | 2089 | Missense Mutation | AAA,AGA | K,R 472 | XP_016882139.1 | |
XM_017026651.1 | 2089 | Missense Mutation | AAA,AGA | K,R 472 | XP_016882140.1 |