Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAACACTCCACACTGTCCAGGAA[C/T]GTGTGACCATCATAGCCTCCTGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606016 | ||||||||||||||||||||
Literature Links: |
KEAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KEAP1 - kelch like ECH associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012289.3 | 2052 | Silent Mutation | ACA,ACG | T,T 576 | NP_036421.2 | |
NM_203500.1 | 2052 | Silent Mutation | ACA,ACG | T,T 576 | NP_987096.1 | |
XM_005260173.1 | 2052 | Silent Mutation | ACA,ACG | T,T 576 | XP_005260230.1 | |
XM_005260174.1 | 2052 | Silent Mutation | ACA,ACG | T,T 576 | XP_005260231.1 | |
XM_011528452.1 | 2052 | Silent Mutation | ACA,ACG | T,T 576 | XP_011526754.1 |