Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGTAGGCCTAAAACCCACATTT[C/T]TAAGAACTATGGGAATAATTTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF845 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF845 - zinc finger protein 845 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321522.1 | 807 | Missense Mutation | TCT,TTT | S,F 189 | NP_001308451.1 | |
NM_001321523.1 | 807 | Missense Mutation | TCT,TTT | S,F 189 | NP_001308452.1 | |
NM_001321524.1 | 807 | Missense Mutation | TCT,TTT | S,F 189 | NP_001308453.1 | |
NM_138374.2 | 807 | Missense Mutation | TCT,TTT | S,F 189 | NP_612383.1 |