Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGGGCCACCCCACCTGGCTCC[A/G]TCATCATCTCGCCACCCAACCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601263 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP2K2 - mitogen-activated protein kinase kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030662.3 | Intron | NP_109587.1 | ||||
XM_006722799.2 | Intron | XP_006722862.1 | ||||
XM_017026989.1 | Intron | XP_016882478.1 | ||||
XM_017026990.1 | Intron | XP_016882479.1 | ||||
XM_017026991.1 | Intron | XP_016882480.1 |