Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGACAACAGAACTCTGCTCCTGG[A/G]AACTCTCCCTTTGCCGAAGGTTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602652 | ||||||||||||||||||||
Literature Links: |
KLK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLK6 - kallikrein related peptidase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012964.2 | 689 | Missense Mutation | TCC,TTC | S,F 83 | NP_001012982.1 | |
NM_001012965.2 | 689 | UTR 5 | NP_001012983.1 | |||
NM_001319948.1 | 689 | UTR 5 | NP_001306877.1 | |||
NM_001319949.1 | 689 | UTR 5 | NP_001306878.1 | |||
NM_002774.3 | 689 | Missense Mutation | TCC,TTC | S,F 83 | NP_002765.1 |