Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGGCAACTTCAGCCTCAGCCCA[C/T]CCTCGCCTGTGGCCGCCTCGCCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611712 MIM: 605725 | ||||||||||||||||||||
Literature Links: |
HIPK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIPK4 - homeodomain interacting protein kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRX - periaxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020956.2 | 4640 | UTR 3 | NP_066007.1 | |||
NM_181882.2 | 4640 | Missense Mutation | NP_870998.2 | |||
XM_011527171.2 | 4640 | Missense Mutation | XP_011525473.1 | |||
XM_017027046.1 | 4640 | Missense Mutation | XP_016882535.1 | |||
XM_017027047.1 | 4640 | Missense Mutation | XP_016882536.1 |