Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCAGAAGATATGAAGCAGGACC[A/G]GGACATTCAGGTGAGTTGGTGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609336 MIM: 616808 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPTL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGPTL6 - angiopoietin like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C19orf66 - chromosome 19 open reading frame 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308277.1 | 533 | Missense Mutation | CAG,CGG | Q,R 75 | NP_001295206.1 | |
NM_018381.3 | 533 | Missense Mutation | CAG,CGG | Q,R 75 | NP_060851.2 | |
XM_011528121.1 | 533 | Missense Mutation | CAG,CGG | Q,R 36 | XP_011526423.1 | |
XM_011528122.1 | 533 | Intron | XP_011526424.1 | |||
XM_017026934.1 | 533 | Missense Mutation | CAG,CGG | Q,R 24 | XP_016882423.1 |