Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGTATCCAGTGGAGCCCAGGTAG[C/G]TGCTGGCGCTGACGGGTCTGAAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615038 MIM: 603996 | ||||||||||||||||||||
Literature Links: |
CCDC114 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC114 - coiled-coil domain containing 114 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144577.3 | 1361 | Missense Mutation | ACC,AGC | T,S 608 | NP_653178.3 | |
XM_005259413.3 | 1361 | Missense Mutation | ACC,AGC | T,S 645 | XP_005259470.1 | |
XM_005259414.3 | 1361 | Missense Mutation | ACC,AGC | T,S 645 | XP_005259471.1 | |
XM_005259415.3 | 1361 | Missense Mutation | ACC,AGC | T,S 645 | XP_005259472.1 | |
XM_005259416.4 | 1361 | Missense Mutation | ACC,AGC | T,S 417 | XP_005259473.1 | |
XM_011527515.2 | 1361 | Missense Mutation | ACC,AGC | T,S 608 | XP_011525817.1 | |
XM_011527516.2 | 1361 | Missense Mutation | ACC,AGC | T,S 608 | XP_011525818.1 | |
XM_011527517.1 | 1361 | Intron | XP_011525819.1 | |||
XM_017027483.1 | 1361 | Missense Mutation | ACC,AGC | T,S 553 | XP_016882972.1 |
ZNF114 - zinc finger protein 114 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |