Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGAAGTTTGATGGGACAAGACT[C/T]TATGAAAGCACAAAGGATGGGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 107266 | ||||||||||||||||||||
Literature Links: |
CD22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD22 - CD22 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185099.1 | 312 | Silent Mutation | CTC,CTT | L,L 76 | NP_001172028.1 | |
NM_001185100.1 | 312 | Silent Mutation | CTC,CTT | L,L 76 | NP_001172029.1 | |
NM_001185101.1 | 312 | Silent Mutation | CTC,CTT | L,L 76 | NP_001172030.1 | |
NM_001278417.1 | 312 | UTR 5 | NP_001265346.1 | |||
NM_001771.3 | 312 | Silent Mutation | CTC,CTT | L,L 76 | NP_001762.2 |