Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTTCAACCGAGGCCCTGCCTA[C/T]GGGCTGTCAGCCGAGGTTAAGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600806 MIM: 608388 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNN1 - calponin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001299.5 | 247 | Silent Mutation | TAC,TAT | Y,Y 12 | NP_001290.2 | |
NM_001308341.1 | 247 | UTR 5 | NP_001295270.1 | |||
NM_001308342.1 | 247 | UTR 5 | NP_001295271.1 | |||
XM_005259741.2 | 247 | Intron | XP_005259798.1 | |||
XM_017026289.1 | 247 | Intron | XP_016881778.1 |
ECSIT - ECSIT signalling integrator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |