Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGTTCATGGTTCCCACCTCGGT[A/G]AGCTGGTGGTCGGGCCCATACAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAT2 - branched chain amino acid transaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164773.1 | 1323 | Silent Mutation | CTC,CTT | L,L 170 | NP_001158245.1 | |
NM_001190.3 | 1323 | Silent Mutation | CTC,CTT | L,L 262 | NP_001181.2 | |
NM_001284325.1 | 1323 | Silent Mutation | CTC,CTT | L,L 222 | NP_001271254.1 |