Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAGCCCACTGGGGTCACAGCCT[A/G]GTGGCGGTAAGGCCCCGTATACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600661 | ||||||||||||||||||||
Literature Links: |
BORCS8-MEF2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BORCS8-MEF2B - BORCS8-MEF2B readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005919.3 | 1176 | Missense Mutation | CCA,CTA | P,L 150 | NP_005910.1 |
MEF2B - myocyte enhancer factor 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145785.1 | 1176 | Missense Mutation | CCA,CTA | P,L 150 | NP_001139257.1 |
TMEM161A - transmembrane protein 161A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |