Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCCGGGAGCTGCTGGTGGCCC[A/G]TCCGCAGGGTGACTGCTGGCTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFSF14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFSF14 - tumor necrosis factor superfamily member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003807.4 | 947 | Missense Mutation | CGG,TGG | R,W 189 | NP_003798.2 | |
NM_172014.3 | 947 | Missense Mutation | CGG,TGG | R,W 153 | NP_742011.2 | |
XM_017027417.1 | 947 | Missense Mutation | CGG,TGG | R,W 189 | XP_016882906.1 | |
XM_017027418.1 | 947 | Intron | XP_016882907.1 |