Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAAGGACCACCAGGATCAAGCC[A/G]GCCATGCCCATGCGGATGAGATTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606047 | ||||||||||||||||||||
Literature Links: |
LILRA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LILRA5 - leukocyte immunoglobulin like receptor A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021250.3 | Intron | NP_067073.1 | ||||
NM_181879.2 | Intron | NP_870994.1 | ||||
NM_181985.3 | Intron | NP_871714.1 | ||||
NM_181986.2 | Intron | NP_871715.1 |