Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGAAGCTGCCTCAGACAGGGC[A/G]CTGGTGCTTAGCTCAGCATAGGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSTM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VSTM1 - V-set and transmembrane domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288791.1 | 1119 | Silent Mutation | AGC,AGT | S,S 124 | NP_001275720.1 | |
NM_001288792.1 | 1119 | Silent Mutation | AGC,AGT | S,S 181 | NP_001275721.1 | |
NM_001288793.1 | 1119 | Silent Mutation | AGC,AGT | S,S 92 | NP_001275722.1 | |
NM_198481.3 | 1119 | Silent Mutation | AGC,AGT | S,S 212 | NP_940883.2 | |
XM_011526845.1 | 1119 | Silent Mutation | AGC,AGT | S,S 340 | XP_011525147.1 | |
XM_011526846.1 | 1119 | Silent Mutation | AGC,AGT | S,S 310 | XP_011525148.1 | |
XM_011526847.1 | 1119 | UTR 3 | XP_011525149.1 | |||
XM_011526848.1 | 1119 | Silent Mutation | AGC,AGT | S,S 279 | XP_011525150.1 | |
XM_011526849.1 | 1119 | Silent Mutation | AGC,AGT | S,S 242 | XP_011525151.1 | |
XM_017026666.1 | 1119 | Intron | XP_016882155.1 |