Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGAGCAGATCATGAAGACAGG[A/G]GCCCTTTTGCTTCAGGGGTGAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600040 MIM: 134790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAX - BCL2 associated X, apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291428.1 | 138 | Silent Mutation | GGA,GGG | G,G 23 | NP_001278357.1 | |
NM_001291429.1 | 138 | Intron | NP_001278358.1 | |||
NM_001291430.1 | 138 | Intron | NP_001278359.1 | |||
NM_001291431.1 | 138 | Intron | NP_001278360.1 | |||
NM_004324.3 | 138 | Silent Mutation | GGA,GGG | G,G 23 | NP_004315.1 | |
NM_138761.3 | 138 | Silent Mutation | GGA,GGG | G,G 23 | NP_620116.1 | |
NM_138763.3 | 138 | Silent Mutation | GGA,GGG | G,G 23 | NP_620118.1 | |
NM_138764.4 | 138 | Silent Mutation | GGA,GGG | G,G 23 | NP_620119.2 | |
XM_017027077.1 | 138 | Silent Mutation | GGA,GGG | G,G 4 | XP_016882566.1 |
FTL - ferritin light chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |