Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGGGATATCTTAAGGGTCCAG[G/T]GACATGGAGGGGGCACTTTGTGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611112 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DACT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DACT3 - dishevelled binding antagonist of beta catenin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301046.1 | 3198 | UTR 3 | NP_001287975.1 | |||
NM_145056.2 | 3198 | UTR 3 | NP_659493.2 | |||
XM_011526499.2 | 3198 | UTR 3 | XP_011524801.1 | |||
XM_011526500.2 | 3198 | UTR 3 | XP_011524802.1 | |||
XM_017026334.1 | 3198 | UTR 3 | XP_016881823.1 |
GNG8 - G protein subunit gamma 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |