Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCTCTTCATCAGATGGGTCTA[A/G]GGATTCAGATGCTATGTCACTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605866 MIM: 611294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP8B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP8B3 - ATPase phospholipid transporting 8B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178002.2 | 3909 | Silent Mutation | CTA,TTA | L,L 1248 | NP_001171473.1 | |
NM_138813.3 | 3909 | Silent Mutation | CTA,TTA | L,L 1285 | NP_620168.1 | |
XM_006722654.2 | 3909 | Silent Mutation | CTA,TTA | L,L 1295 | XP_006722717.1 | |
XM_006722655.2 | 3909 | Silent Mutation | CTA,TTA | L,L 1295 | XP_006722718.1 | |
XM_006722656.2 | 3909 | Silent Mutation | CTA,TTA | L,L 1209 | XP_006722719.1 | |
XM_011527707.1 | 3909 | Silent Mutation | CTA,TTA | L,L 1286 | XP_011526009.1 | |
XM_011527708.1 | 3909 | Silent Mutation | CTA,TTA | L,L 1255 | XP_011526010.1 | |
XM_011527709.1 | 3909 | Silent Mutation | CTA,TTA | L,L 1224 | XP_011526011.1 | |
XM_011527710.2 | 3909 | Silent Mutation | CTA,TTA | L,L 1043 | XP_011526012.1 | |
XM_011527711.2 | 3909 | Intron | XP_011526013.1 | |||
XM_011527712.1 | 3909 | Silent Mutation | CTA,TTA | L,L 675 | XP_011526014.1 |
ONECUT3 - one cut homeobox 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |