Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGAGAGATGGCTAATGAATGG[C/T]GCTTGATGACAGATGAGTAATGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF559 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF559 - zinc finger protein 559 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202406.1 | 563 | Intron | NP_001189335.1 | |||
NM_001202407.1 | 563 | Intron | NP_001189336.1 | |||
NM_001202408.1 | 563 | Intron | NP_001189337.1 | |||
NM_001202409.1 | 563 | UTR 5 | NP_001189338.1 | |||
NM_001202410.1 | 563 | UTR 5 | NP_001189339.1 | |||
NM_001202411.1 | 563 | Intron | NP_001189340.1 | |||
NM_001202412.1 | 563 | UTR 5 | NP_001189341.1 | |||
NM_032497.2 | 563 | UTR 5 | NP_115886.1 |
ZNF559-ZNF177 - ZNF559-ZNF177 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172650.2 | 563 | Intron | NP_001166121.1 | |||
NM_001202425.1 | 563 | Intron | NP_001189354.1 |