Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTCTGAGCTGCATCTGAAGGCC[C/T]TTCCACATGCCATGCAATCATAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF549 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF549 - zinc finger protein 549 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF550 - zinc finger protein 550 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277090.1 | 1227 | Missense Mutation | AAG,AGG | K,R 350 | NP_001264019.1 | |
NM_001277091.1 | 1227 | Missense Mutation | AAG,AGG | K,R 350 | NP_001264020.1 | |
NM_001277092.1 | 1227 | Missense Mutation | AAG,AGG | K,R 350 | NP_001264021.1 | |
NM_001277093.1 | 1227 | Intron | NP_001264022.1 | |||
XM_011526567.2 | 1227 | Missense Mutation | AAG,AGG | K,R 350 | XP_011524869.1 | |
XM_011526568.2 | 1227 | Missense Mutation | AAG,AGG | K,R 341 | XP_011524870.1 | |
XM_017026401.1 | 1227 | Missense Mutation | AAG,AGG | K,R 350 | XP_016881890.1 |