Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTCAGAAGGTCGCCATCAAGAT[C/T]GTGAACCGGGAGAAGCTGTCGGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609235 MIM: 612939 | ||||||||||||||||||||
Literature Links: |
BRSK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRSK1 - BR serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032430.1 | 469 | Silent Mutation | ATC,ATT | I,I 64 | NP_115806.1 | |
XM_005259327.3 | 469 | UTR 5 | XP_005259384.1 | |||
XM_011527395.2 | 469 | Intron | XP_011525697.2 |
HSPBP1 - HSPA (Hsp70) binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |