Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAACTGACAGCAACGATGGACAT[A/G]GGTGCGCTGAAGCCAATGATGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136537 MIM: 136538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FPR1 - formyl peptide receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193306.1 | 734 | Silent Mutation | CCC,CCT | P,P 213 | NP_001180235.1 | |
NM_002029.3 | 734 | Silent Mutation | CCC,CCT | P,P 213 | NP_002020.1 |
FPR2 - formyl peptide receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |