Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAACAGGTGCGAGGGTCTGGGG[A/G]CTCCGCTCCTCCTTCAAGGTCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607548 MIM: 602388 | ||||||||||||||||||||
Literature Links: |
RSPH6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSPH6A - radial spoke head 6 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030785.3 | 3927 | Intron | NP_110412.1 | |||
XM_011527351.1 | 3927 | Intron | XP_011525653.1 |
SYMPK - symplekin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004819.2 | 3927 | Silent Mutation | AGC,AGT | S,S 1243 | NP_004810.2 | |
XM_005259286.1 | 3927 | Silent Mutation | AGC,AGT | S,S 1243 | XP_005259343.1 | |
XM_011527354.1 | 3927 | Silent Mutation | AGC,AGT | S,S 1243 | XP_011525656.1 |