Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCGCATACCTCCTTCACGTTGG[C/T]AACCTGTGCCTGGTGATGGGATGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HSPC102 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPC102 - uncharacterized HSPC102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN16 - tetraspanin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282509.1 | 264 | Silent Mutation | GGC,GGT | G,G 58 | NP_001269438.1 | |
NM_001282510.1 | 264 | Silent Mutation | GGC,GGT | G,G 58 | NP_001269439.1 | |
NM_012466.3 | 264 | Silent Mutation | GGC,GGT | G,G 58 | NP_036598.1 | |
XM_011527901.1 | 264 | Silent Mutation | GGC,GGT | G,G 58 | XP_011526203.1 | |
XM_011527902.1 | 264 | Silent Mutation | GGC,GGT | G,G 58 | XP_011526204.1 | |
XM_011527903.2 | 264 | UTR 5 | XP_011526205.1 |