Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTTCCACATTCATGACATTCA[C/T]GAGATCTTACTCCAGTATGAACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606956 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF256 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF256 - zinc finger protein 256 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005773.2 | 1539 | Missense Mutation | CAT,CGT | H,R 435 | NP_005764.2 | |
XM_017026138.1 | 1539 | Missense Mutation | CAT,CGT | H,R 282 | XP_016881627.1 |
ZNF418 - zinc finger protein 418 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |