Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCGGACTGGATGGCTGTGGCC[A/G]CTTTGTCCTGGCGATGCGGCTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1683 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1683 - KIAA1683 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145304.1 | 4124 | Missense Mutation | GCG,GTG | A,V 1303 | NP_001138776.1 | |
NM_001145305.1 | 4124 | Missense Mutation | GCG,GTG | A,V 1070 | NP_001138777.1 | |
NM_025249.3 | 4124 | Missense Mutation | GCG,GTG | A,V 1116 | NP_079525.1 | |
XM_005260084.1 | 4124 | Missense Mutation | GCG,GTG | A,V 1303 | XP_005260141.1 | |
XM_011528324.1 | 4124 | Missense Mutation | GCG,GTG | A,V 1257 | XP_011526626.1 |
LOC729966 - uncharacterized LOC729966 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE4C - phosphodiesterase 4C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |