Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGAAGGAATCCACTCAACAGAC[A/G]AGGTACCTTAACCACATAATGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF528 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF528 - zinc finger protein 528 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032423.2 | 349 | UTR 5 | NP_115799.2 | |||
XM_005259323.1 | 349 | Intron | XP_005259380.1 | |||
XM_006723418.2 | 349 | UTR 5 | XP_006723481.1 | |||
XM_011527392.1 | 349 | UTR 5 | XP_011525694.1 | |||
XM_011527394.2 | 349 | Intron | XP_011525696.1 | |||
XM_017027360.1 | 349 | UTR 5 | XP_016882849.1 | |||
XM_017027361.1 | 349 | UTR 5 | XP_016882850.1 | |||
XM_017027362.1 | 349 | UTR 5 | XP_016882851.1 | |||
XM_017027363.1 | 349 | UTR 5 | XP_016882852.1 | |||
XM_017027364.1 | 349 | UTR 5 | XP_016882853.1 | |||
XM_017027365.1 | 349 | Intron | XP_016882854.1 |
ZNF528-AS1 - ZNF528 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF880 - zinc finger protein 880 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |