Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTCGCCGACCTGCGGGCCCTGC[G/T]TGCCCTGCACCTGGATGGCAACCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612809 | ||||||||||||||||||||
Literature Links: |
LOC105372383 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105372383 - uncharacterized LOC105372383 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRFN3 - leucine rich repeat and fibronectin type III domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024509.1 | 781 | Missense Mutation | CGT,CTT | R,L 110 | NP_078785.1 | |
XM_017027302.1 | 781 | Missense Mutation | CGT,CTT | R,L 110 | XP_016882791.1 |