Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACACCATCGACACCGAAAACTC[C/T]ACCGTAGCCCTTGCCAAAGGTACGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610677 | ||||||||||||||||||||
Literature Links: |
LSM14A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LSM14A - LSM14A, mRNA processing body assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114093.1 | 317 | Silent Mutation | TCC,TCT | S,S 34 | NP_001107565.1 | |
NM_015578.2 | 317 | Silent Mutation | TCC,TCT | S,S 34 | NP_056393.2 | |
XM_005258719.4 | 317 | Silent Mutation | TCC,TCT | S,S 34 | XP_005258776.1 | |
XM_005258720.4 | 317 | Silent Mutation | TCC,TCT | S,S 34 | XP_005258777.1 | |
XM_005258721.4 | 317 | Silent Mutation | TCC,TCT | S,S 34 | XP_005258778.1 | |
XM_011526708.2 | 317 | Silent Mutation | TCC,TCT | S,S 34 | XP_011525010.1 | |
XM_017026575.1 | 317 | Silent Mutation | TCC,TCT | S,S 34 | XP_016882064.1 | |
XM_017026576.1 | 317 | Silent Mutation | TCC,TCT | S,S 34 | XP_016882065.1 | |
XM_017026577.1 | 317 | Silent Mutation | TCC,TCT | S,S 34 | XP_016882066.1 |