Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCTTCTCATGTGCCGTTCCACA[A/G]GGCACCTGCTCTTCCAGCAGCTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604692 | ||||||||||||||||||||
Literature Links: |
AKAP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP8 - A-kinase anchoring protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005858.3 | 2053 | Silent Mutation | CCC,CCT | P,P 630 | NP_005849.1 | |
XM_011527624.2 | 2053 | Silent Mutation | CCC,CCT | P,P 530 | XP_011525926.1 | |
XM_017026141.1 | 2053 | Silent Mutation | CCC,CCT | P,P 556 | XP_016881630.1 |