Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGCTCGTGGCGTGCGACAACGC[A/G]GCGCAGAAGTAAGTAGCCGGGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604948 | ||||||||||||||||||||
Literature Links: |
GADD45B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GADD45B - growth arrest and DNA damage inducible beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015675.3 | 270 | Silent Mutation | GCA,GCG | A,A 12 | NP_056490.2 | |
XM_017026822.1 | 270 | Silent Mutation | GCA,GCG | A,A 12 | XP_016882311.1 |