Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGGCCCCCACGATGATGGGGCA[C/T]ACCATGCCCGACAGTGTGCCCACGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608386 MIM: 605208 | ||||||||||||||||||||
Literature Links: |
GFY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GFY - golgi associated olfactory signaling regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTH2 - parathyroid hormone 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC17A7 - solute carrier family 17 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020309.3 | 1525 | Silent Mutation | GTA,GTG | V,V 451 | NP_064705.1 |